Assay Details
Gene Symbol: Phactr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214457.2 | NP_999622.2 | 11 - 12 | 1701 | 145 | |
GenBank mRNA | AY494977.1 | - | 1570 | 145 | ||
BC098634.1 | - | 11 - 12 | 1701 | 7317963 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
