Assay Details
Gene Symbol: Hivep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105751.1 | NP_001099221.1 | 6 - 7 | 6291 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
