Assay Details
Gene Symbol: Bphl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037206.1 | NP_001032283.1 | 2 - 3 | 219 | 72 | |
GenBank mRNA | BC105908.1 | - | 2 - 3 | 219 | 72 | |
FQ215116.1 | - | 2 - 3 | 229 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
