Assay Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107348.1 | NP_001100818.1 | 3 - 4 | 374 | 133 | |
XM_006253812.2 | XP_006253874.1 | 2 - 3 | 352 | 133 | ||
XM_006253813.2 | XP_006253875.1 | 1 - 2 | 106 | 133 | ||
XM_006253814.2 | XP_006253876.1 | 2 - 3 | 721 | 133 | ||
XM_006253815.2 | XP_006253877.1 | 2 - 3 | 1318 | 133 | ||
XM_006253816.2 | XP_006253878.1 | 2 - 3 | 352 | 133 | ||
XM_008771571.1 | XP_008769793.1 | 1 - 2 | 106 | 133 | ||
GenBank mRNA | FQ222841.1 | - | 2 - 3 | 167 | 133 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001107348.1, XM_006253812.2, XM_006253813.2, XM_006253814.2, XM_006253815.2, XM_006253816.2, XM_008771571.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
