Assay Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107348.1 | NP_001100818.1 | 7 - 8 | 954 | 77 | |
XM_006253812.2 | XP_006253874.1 | 6 - 7 | 932 | 77 | ||
XM_006253813.2 | XP_006253875.1 | 5 - 6 | 686 | 77 | ||
XM_006253814.2 | XP_006253876.1 | 6 - 7 | 1301 | 77 | ||
XM_006253815.2 | XP_006253877.1 | 6 - 7 | 1898 | 77 | ||
XM_006253816.2 | XP_006253878.1 | 6 - 7 | 932 | 77 | ||
XM_006253817.2 | XP_006253879.1 | 3 - 4 | 1431 | 77 | ||
XM_008771571.1 | XP_008769793.1 | 5 - 6 | 686 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
