Assay Details
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107348.1 | NP_001100818.1 | 8 - 9 | 1096 | 73 | |
XM_006253812.2 | XP_006253874.1 | 7 - 8 | 1074 | 73 | ||
XM_006253813.2 | XP_006253875.1 | 6 - 7 | 828 | 73 | ||
XM_006253814.2 | XP_006253876.1 | 7 - 8 | 1443 | 73 | ||
XM_006253815.2 | XP_006253877.1 | 7 - 8 | 2040 | 73 | ||
XM_006253816.2 | XP_006253878.1 | 7 - 8 | 1074 | 73 | ||
XM_006253817.2 | XP_006253879.1 | 4 - 5 | 1573 | 73 | ||
XM_008771571.1 | XP_008769793.1 | 6 - 7 | 828 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
