Assay Details
Gene Symbol: Slc35b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751664.3 | XP_003751712.1 | 2 - 3 | 406 | 111 | |
XM_006253850.1 | XP_006253912.1 | 2 - 3 | 386 | 111 | ||
XM_006253852.2 | XP_006253914.1 | 2 - 3 | 546 | 111 | ||
XM_006253853.2 | XP_006253915.1 | 2 - 3 | 546 | 111 | ||
XM_006253854.2 | XP_006253916.1 | 2 - 3 | 546 | 111 | ||
XM_006253855.2 | XP_006253917.1 | 2 - 3 | 461 | 111 | ||
XM_006253856.2 | XP_006253918.1 | 2 - 3 | 393 | 111 | ||
XM_225253.8 | XP_225253.5 | 2 - 3 | 545 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
