Assay Details
Gene Symbol: Pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037356.1 | NP_001032433.1 | 1 - 2 | 139 | 64 | |
GenBank mRNA | BC079236.1 | - | 1 - 2 | 119 | 64 | |
BC089822.1 | - | 1 - 2 | 112 | 64 | ||
BC107922.1 | - | 1 - 2 | 139 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
