Assay Details
Gene Symbol: Ranbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771586.1 | XP_008769808.1 | 6 - 7 | 1002 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
