Assay Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024871.1 | NP_001020042.1 | 3 - 4 | 483 | 74 | |
XM_006233955.1 | XP_006234017.1 | 2 - 3 | 530 | 74 | ||
XM_006233957.2 | XP_006234019.1 | 2 - 3 | 138 | 74 | ||
XM_006233958.1 | XP_006234020.1 | 2 - 3 | 397 | 74 | ||
XM_006233959.2 | XP_006234021.1 | 2 - 3 | 208 | 74 | ||
XM_006233966.1 | XP_006234028.1 | 2 - 3 | 530 | 74 | ||
XM_006233967.1 | XP_006234029.1 | 2 - 3 | 397 | 74 | ||
XM_008761726.1 | XP_008759948.1 | 3 - 4 | 211 | 74 | ||
XM_008761737.1 | XP_008759959.1 | 3 - 4 | 211 | 74 | ||
GenBank mRNA | BC097415.1 | - | 3 - 4 | 483 | 74 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_006233966.1, XM_006233967.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
