Assay Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006234049.2 | XP_006234111.1 | 2 - 3 | 490 | 118 | |
XM_006234050.2 | XP_006234112.1 | 2 - 3 | 142 | 118 | ||
GenBank mRNA | BC158585.1 | - | 1 - 2 | 646 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
