Assay Details
Gene Symbol: Rab14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053589.1 | NP_446041.1 | 5 - 6 | 443 | 126 | |
XM_006234049.2 | XP_006234111.1 | 6 - 7 | 876 | 126 | ||
XM_006234050.2 | XP_006234112.1 | 6 - 7 | 528 | 126 | ||
GenBank mRNA | BC158585.1 | - | 5 - 6 | 1032 | 126 | |
M83680.1 | - | 5 - 6 | 443 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
