Assay Details
Gene Symbol: Ntng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107825.1 | NP_001101295.1 | 1 - 2 | 449 | 136 | |
XM_006233800.2 | XP_006233862.1 | 1 - 2 | 449 | 136 | ||
XM_006233804.2 | XP_006233866.1 | 1 - 2 | 449 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
