Assay Details
Gene Symbol: Ntng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107825.1 | NP_001101295.1 | 2 - 3 | 1087 | 111 | |
XM_006233800.2 | XP_006233862.1 | 2 - 3 | 1087 | 111 | ||
XM_006233802.2 | XP_006233864.1 | 2 - 3 | 1396 | 111 | ||
XM_006233804.2 | XP_006233866.1 | 2 - 3 | 1087 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
