Assay Details
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009692.1 | NP_001009692.1 | 1 - 2 | 116 | 118 | |
XM_006233816.2 | XP_006233878.1 | 1 - 2 | 139 | 118 | ||
XM_006233817.2 | XP_006233879.1 | 1 - 2 | 139 | 118 | ||
XM_006233818.2 | XP_006233880.1 | 1 - 2 | 138 | 118 | ||
XM_006233819.2 | XP_006233881.1 | 1 - 2 | 138 | 118 | ||
XM_006233820.2 | XP_006233882.1 | 1 - 2 | 138 | 118 | ||
XM_006233822.2 | XP_006233884.1 | 1 - 2 | 138 | 118 | ||
XM_008761661.1 | XP_008759883.1 | 1 - 2 | 139 | 118 | ||
XM_008761662.1 | XP_008759884.1 | 1 - 2 | 138 | 118 | ||
GenBank mRNA | BC087652.1 | - | 1 - 2 | 116 | 7312960 | 118 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC087652.1,NM_001009692.1,XM_006233816.2,XM_006233817.2,XM_006233818.2,XM_006233819.2,XM_006233820.2,XM_006233822.2,XM_008761661.1,XM_008761662.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
