Assay Details
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009692.1 | NP_001009692.1 | 7 - 8 | 739 | 126 | |
XM_006233818.2 | XP_006233880.1 | 7 - 8 | 715 | 126 | ||
XM_006233819.2 | XP_006233881.1 | 6 - 7 | 703 | 126 | ||
XM_006233820.2 | XP_006233882.1 | 6 - 7 | 703 | 126 | ||
GenBank mRNA | BC087652.1 | - | 7 - 8 | 739 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
