Assay Details
Gene Symbol: Uck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107831.3 | NP_001101301.2 | 6 - 7 | 737 | 77 | |
GenBank mRNA | BC168897.1 | - | 6 - 7 | 676 | 8362491 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
