Assay Details
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033565.1 | NP_001028737.1 | 3 - 4 | 404 | 64 | |
XM_006233639.2 | XP_006233701.1 | 4 - 5 | 676 | 64 | ||
XM_006233640.2 | XP_006233702.1 | 5 - 6 | 770 | 64 | ||
XM_006233642.2 | XP_006233704.1 | 4 - 5 | 661 | 64 | ||
XM_006233643.2 | XP_006233705.1 | 4 - 5 | 676 | 64 | ||
GenBank mRNA | AY536920.1 | - | 3 - 4 | 404 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
