Assay Details
Gene Symbol: Entpd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033565.1 | NP_001028737.1 | 8 - 9 | 1303 | 58 | |
XM_006233639.2 | XP_006233701.1 | 9 - 10 | 1575 | 58 | ||
XM_006233640.2 | XP_006233702.1 | 10 - 11 | 1669 | 58 | ||
XM_006233642.2 | XP_006233704.1 | 9 - 10 | 1560 | 58 | ||
XM_006233643.2 | XP_006233705.1 | 9 - 10 | 1545 | 58 | ||
GenBank mRNA | AY536920.1 | - | 8 - 9 | 1303 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
