Assay Details
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238127.2 | XP_006238189.1 | 4 - 5 | 858 | 74 | |
XM_233141.8 | XP_233141.7 | 4 - 5 | 1401 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
