Assay Details
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238127.2 | XP_006238189.1 | 7 - 8 | 2755 | 71 | |
XM_233141.8 | XP_233141.7 | 7 - 8 | 3298 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
