Assay Details
Gene Symbol: Ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012960.2 | NP_037092.1 | 4 - 5 | 509 | 101 | |
GenBank mRNA | BC087602.1 | - | 4 - 5 | 509 | 101 | |
U38379.1 | - | 4 - 5 | 359 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
