Assay Details
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113754.1 | NP_001107226.1 | 2 - 3 | 275 | 61 | |
GenBank mRNA | BC158680.1 | - | 2 - 3 | 259 | 61 | |
FQ210026.1 | - | 2 - 3 | 246 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
