Assay Details
Gene Symbol: Grhpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113754.1 | NP_001107226.1 | 5 - 6 | 548 | 64 | |
GenBank mRNA | BC158680.1 | - | 5 - 6 | 532 | 8373346 | 64 |
FQ210026.1 | - | 5 - 6 | 519 | 64 | ||
FQ210631.1 | - | 5 - 6 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
