Assay Details
Gene Symbol: Polr1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107938.1 | NP_001101408.1 | 10 - 11 | 1033 | 97 | |
XM_008763673.1 | XP_008761895.1 | 10 - 11 | 1365 | 97 | ||
GenBank mRNA | BC158699.1 | - | 10 - 11 | 1021 | 9028317 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
