Assay Details
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135780.1 | NP_001129252.1 | 4 - 5 | 820 | 101 | |
XM_006237954.2 | XP_006238016.1 | 4 - 5 | 872 | 101 | ||
GenBank mRNA | BC160888.1 | - | 4 - 5 | 777 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
