Assay Details
Gene Symbol: Mms22l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135780.1 | NP_001129252.1 | 21 - 22 | 3772 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P397198 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P010214 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P156947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12197 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156947 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P397198 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010214 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P010214 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P397198 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P156947 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P397198 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P010214 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P156947 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867923 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867928 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867934 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867939 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867937 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867930 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867938 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867933 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867925 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867920 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867921 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867927 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867936 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867935 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867926 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867931 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867924 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867940 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867932 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867922 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867929 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867919 | - | - |
Rat Genome 230 2.0 Array | 1380243_at | - | - |
Rat Genome 230 2.0 Array | 1390321_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390321_at | - | - |
Rat Genome 230 2.0 Array | 1380243_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390321_at | - | - |
Affymetrix HT Rat Focus Array | 1380243_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380243_at | - | - |
Affymetrix HT Rat Focus Array | 1390321_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500002083.rn.1 | TC0500000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002084.rn.1 | TC0500000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002081.rn.1 | TC0500000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002082.rn.1 | TC0500000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002085.rn.1 | TC0500000330.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002080.rn.1 | TC0500000330.rn.1 | RTA TC |
Rat Expression Set 230 | 1390321_at | - | - |
Rat Expression Set 230 | 1390321_PM_at | - | - |
Rat Expression Set 230 | 1380243_PM_at | - | - |
Rat Expression Set 230 | 1380243_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867919 | - | - |
Rat Genome U34 Set | rc_AA996953_at | - | - |
Rat Genome U34 Set | rc_AI228158_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22290520 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350397 | - | - |
