Assay Details
Gene Symbol: Ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012190.1 | NP_001012190.1 | 5 - 6 | 1471 | 121 | |
GenBank mRNA | BC083594.1 | - | 5 - 6 | 1471 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P557979 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P308197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424162 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153375 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P557979 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P308197 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P308197 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P557979 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P557979 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P308197 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P557979 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P308197 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26555 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88044 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77522 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10868355 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868352 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868350 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868351 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868354 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868353 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10868349 | - | - |
Rat Genome 230 2.0 Array | 1388677_at | - | - |
Rat Genome 230 2.0 Array | 1388677_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388677_at | - | - |
Affymetrix HT Rat Focus Array | 1388677_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500002945.rn.1 | TC0500000487.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500002944.rn.1 | TC0500000487.rn.1 | RTA TC |
Rat Expression Set 230 | 1388677_at | - | - |
Rat Expression Set 230 | 1388677_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10868349 | - | - |
Rat Genome U34 Set | rc_AI169374_at | - | - |
Rat Genome U34 Set | AA850279_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21683308 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376487 | - | - |
