Assay Details
Gene Symbol: LOC100910771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750753.3 | XP_003750801.1 | 15 - 16 | 1655 | 64 | |
XM_006245614.2 | XP_006245676.1 | 14 - 15 | 1574 | 64 |
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107920.2 | NP_001101390.2 | 15 - 16 | 1679 | 64 | |
XM_006237966.2 | XP_006238028.1 | 14 - 15 | 2790 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
