Assay Details
Gene Symbol: LOC100910771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750753.3 | XP_003750801.1 | 7 - 8 | 867 | 81 | |
XM_006245614.2 | XP_006245676.1 | 7 - 8 | 867 | 81 |
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107920.2 | NP_001101390.2 | 7 - 8 | 891 | 81 | |
XM_006237966.2 | XP_006238028.1 | 7 - 8 | 2083 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
neural tube formation
positive regulation of T cell cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
transforming growth factor beta receptor signaling pathway
activation of NF-kappaB-inducing kinase activity
I-kappaB phosphorylation
JNK cascade
positive regulation of interleukin-2 production
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
histone H3 acetylation
positive regulation of JNK cascade
stress-activated MAPK cascade
negative regulation of necroptotic process
negative regulation of ripoptosome assembly involved in necroptotic process
regulation of reactive oxygen species metabolic process
negative regulation of reactive oxygen species metabolic process
negative regulation of apoptotic signaling pathway
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
scaffold protein binding
MAPK cascade
angiogenesis
neural tube formation
positive regulation of T cell cytokine production
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
transforming growth factor beta receptor signaling pathway
activation of NF-kappaB-inducing kinase activity
I-kappaB phosphorylation
JNK cascade
positive regulation of interleukin-2 production
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JUN kinase activity
histone H3 acetylation
positive regulation of JNK cascade
stress-activated MAPK cascade
negative regulation of necroptotic process
negative regulation of ripoptosome assembly involved in necroptotic process
regulation of reactive oxygen species metabolic process
negative regulation of reactive oxygen species metabolic process
negative regulation of apoptotic signaling pathway
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
scaffold protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P021723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119471 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P021723 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P021723 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P021723 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P932802 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P300018 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11232 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867985 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867991 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867984 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867987 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867980 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867993 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867989 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867992 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867990 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867986 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867981 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867988 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867979 | - | - |
Rat Genome 230 2.0 Array | 1399075_at | - | - |
Rat Genome 230 2.0 Array | 1399075_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399075_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1399075_at | - | - |
Rat Expression Set 230 | 1399075_PM_at | - | - |
Rat Expression Set 230 | 1399075_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867979 | - | - |
Rat Genome U34 Set | rc_AI146037_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21237053 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371190 | - | - |
