Assay Details
Gene Symbol: Trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013090.2 | NP_001013108.1 | 4 - 5 | 608 | 136 | |
XM_006238042.1 | XP_006238104.1 | 5 - 6 | 658 | 136 | ||
XM_006238043.1 | XP_006238105.1 | 5 - 6 | 857 | 136 | ||
XM_006238044.2 | XP_006238106.1 | 5 - 6 | 618 | 136 | ||
GenBank mRNA | BC086603.1 | - | 4 - 5 | 612 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
