Assay Details
Gene Symbol: Trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013090.2 | NP_001013108.1 | 6 - 7 | 747 | 115 | |
XM_006238042.1 | XP_006238104.1 | 7 - 8 | 797 | 115 | ||
XM_006238043.1 | XP_006238105.1 | 7 - 8 | 996 | 115 | ||
XM_006238044.2 | XP_006238106.1 | 7 - 8 | 757 | 115 | ||
GenBank mRNA | BC086603.1 | - | 6 - 7 | 751 | 7315452 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
