Assay Details
Gene Symbol: Arhgef39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106676.2 | NP_001100146.2 | 5 - 6 | 563 | 65 | |
GenBank mRNA | BC158673.1 | - | 5 - 6 | 563 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
