Assay Details
Gene Symbol: Ccl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238081.1 | XP_006238143.1 | 1 - 2 | 963 | 82 | |
GenBank mRNA | FQ228666.1 | - | 1 - 2 | 98 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
