Assay Details
Gene Symbol: Sigmar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030996.1 | NP_112258.1 | 2 - 3 | 388 | 129 | |
GenBank mRNA | AF004218.1 | - | 2 - 3 | 388 | 129 | |
AF067769.1 | - | 2 - 3 | 440 | 129 | ||
BC061978.1 | - | 2 - 3 | 377 | 5623693 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
