Assay Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053953.1 | NP_446405.1 | 4 - 5 | 666 | 111 | |
XM_008766983.1 | XP_008765205.1 | 4 - 5 | 1051 | 111 | ||
XM_008766984.1 | XP_008765206.1 | 4 - 5 | 822 | 111 | ||
GenBank mRNA | BC091564.1 | - | 4 - 5 | 640 | 7445971 | 111 |
FQ228742.1 | - | 4 - 5 | 665 | 111 | ||
FQ229102.1 | - | 4 - 5 | 665 | 111 | ||
Z22812.1 | - | 4 - 5 | 666 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
