Assay Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766983.1 | XP_008765205.1 | 5 - 6 | 1230 | 119 | |
XM_008766984.1 | XP_008765206.1 | 5 - 6 | 1001 | 119 | ||
GenBank mRNA | BC091564.1 | - | 5 - 6 | 819 | 7445971 | 119 |
FQ228742.1 | - | 5 - 6 | 844 | 119 | ||
FQ229102.1 | - | 5 - 6 | 844 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
