Assay Details
Gene Symbol: Il1r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053953.1 | NP_446405.1 | 7 - 8 | 1040 | 134 | |
XM_008766983.1 | XP_008765205.1 | 7 - 8 | 1425 | 134 | ||
XM_008766984.1 | XP_008765206.1 | 7 - 8 | 1196 | 134 | ||
GenBank mRNA | BC091564.1 | - | 7 - 8 | 1014 | 7445971 | 134 |
FQ228742.1 | - | 7 - 8 | 1039 | 134 | ||
FQ229102.1 | - | 7 - 8 | 1037 | 132 | ||
Z22812.1 | - | 7 - 8 | 1040 | 134 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
