Assay Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100549.1 | NP_001094019.1 | 10 - 11 | 1118 | 123 | |
GenBank mRNA | BC088242.1 | - | 9 - 10 | 930 | 123 | |
BC158704.1 | - | 10 - 11 | 1086 | 123 | ||
BP498271.1 | - | 1 - 2 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P485592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72512 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P485592 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P485592 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P485592 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P485592 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33621 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47259 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10922940 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922948 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922942 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922941 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922938 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922943 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922937 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922947 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922946 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922944 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922939 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10922936 | - | - |
Rat Genome 230 2.0 Array | 1373002_at | - | - |
Rat Genome 230 2.0 Array | 1373002_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396142_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396142_at | - | - |
Affymetrix HT Rat Focus Array | 1373002_at | - | - |
Affymetrix HT Rat Focus Array | 1373002_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396142_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396142_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900004560.rn.1 | TC0900000565.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900004561.rn.1 | TC0900000565.rn.1 | RTA TC |
Rat Expression Set 230 | 1373002_at | - | - |
Rat Expression Set 230 | 1373002_PM_at | - | - |
Rat Expression Set 230 | 1396142_at | - | - |
Rat Expression Set 230 | 1396142_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10922936 | - | - |
Rat Genome U34 Set | rc_AI009481_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21924660 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353438 | - | - |
