Assay Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100549.1 | NP_001094019.1 | 3 - 4 | 414 | 61 | |
XM_008767019.1 | XP_008765241.1 | 3 - 4 | 425 | 61 | ||
GenBank mRNA | BC088242.1 | - | 2 - 3 | 226 | 61 | |
BC158704.1 | - | 3 - 4 | 382 | 61 | ||
CV106852.1 | - | 3 - 4 | 414 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
