Assay Details
Gene Symbol: Mrps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100549.1 | NP_001094019.1 | 8 - 9 | 840 | 102 | |
XM_008767019.1 | XP_008765241.1 | 8 - 9 | 851 | 102 | ||
GenBank mRNA | BC088242.1 | - | 7 - 8 | 652 | 102 | |
BC158704.1 | - | 8 - 9 | 808 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
