Assay Details
Gene Symbol: Tyw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170473.1 | NP_001163944.1 | 4 - 5 | 619 | 63 | |
NR_033170.1 | 4 - 5 | 600 | 63 | |||
GenBank mRNA | BC158619.1 | - | 4 - 5 | 600 | 63 | |
FM112098.1 | - | 4 - 5 | 334 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
