Assay Details
Gene Symbol: Ino80d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191809.1 | NP_001178738.1 | 7 - 8 | 1747 | 65 | |
XM_006245023.2 | XP_006245085.1 | 10 - 11 | 2116 | 65 | ||
XM_006245024.2 | XP_006245086.1 | 9 - 10 | 3449 | 65 | ||
XM_006245025.2 | XP_006245087.1 | 9 - 10 | 2012 | 65 | ||
XM_006245026.2 | XP_006245088.1 | 10 - 11 | 2359 | 65 | ||
XM_006245029.2 | XP_006245091.1 | 10 - 11 | 2226 | 65 | ||
XM_006245031.2 | XP_006245093.1 | 9 - 10 | 2122 | 65 | ||
XM_006245032.2 | XP_006245094.1 | 9 - 10 | 2255 | 65 | ||
XM_006245033.2 | XP_006245095.1 | 9 - 10 | 2031 | 65 | ||
XM_006245034.2 | XP_006245096.1 | 9 - 10 | 1921 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
