Assay Details
Gene Symbol: Rab23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109005.1 | NP_001102475.1 | 2 - 3 | 279 | 69 | |
XM_008766960.1 | XP_008765182.1 | 2 - 3 | 933 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001109005.1, XM_008766960.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
