Assay Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 10 - 11 | 1132 | 91 | |
GenBank mRNA | BC085338.1 | - | 2 - 3 | 81 | 91 | |
BC099152.1 | - | 4 - 5 | 1279 | 91 | ||
BC166803.1 | - | 10 - 11 | 1099 | 7938869 | 91 | |
FQ228979.1 | - | 10 - 11 | 1100 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
