Assay Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 5 - 6 | 540 | 67 | |
GenBank mRNA | BC166803.1 | - | 5 - 6 | 507 | 67 | |
DY315273.1 | - | 2 - 3 | 153 | 67 | ||
EV779654.1 | - | 5 - 6 | 568 | 67 | ||
FQ228979.1 | - | 5 - 6 | 508 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
