Assay Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 8 - 9 | 1032 | 147 | |
GenBank mRNA | BC166803.1 | - | 8 - 9 | 999 | 7938869 | 147 |
FQ228979.1 | - | 8 - 9 | 1000 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
