Assay Details
Gene Symbol: Cnot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108794.2 | NP_001102264.1 | 1 - 2 | 565 | 122 | |
GenBank mRNA | BC158750.1 | - | 1 - 2 | 565 | 122 | |
FQ233266.1 | - | 1 - 2 | 570 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
