Assay Details
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106913.2 | NP_001100383.2 | 6 - 7 | 802 | 97 | |
XM_008767083.1 | XP_008765305.1 | 5 - 6 | 731 | 97 | ||
GenBank mRNA | BC081942.1 | - | 4 - 5 | 2061 | 97 | |
BC097451.1 | - | 4 - 5 | 2018 | 97 | ||
FQ219215.1 | - | 2 - 3 | 375 | 97 | ||
FQ232068.1 | - | 6 - 7 | 812 | 97 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the XM_008767083.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
