Assay Details
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106913.2 | NP_001100383.2 | 9 - 10 | 1182 | 116 | |
XM_008767083.1 | XP_008765305.1 | 8 - 9 | 1111 | 116 | ||
GenBank mRNA | BC081942.1 | - | 7 - 8 | 2441 | 116 | |
BC097451.1 | - | 7 - 8 | 2398 | 116 | ||
FQ219215.1 | - | 5 - 6 | 755 | 116 | ||
FQ232068.1 | - | 9 - 10 | 1192 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
